Benzhydrylamine
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: α-Phenylbenzylamine, α-Aminodiphenylmethane, Diphenylmethanamine, Aminodiphenylmethane, 1,1-Diphenylmethanamine, Benzhydrylamin
- CAS Number:91-00-9
- Formula:C₁₃H₁₃N
- MDL Number:MFCD00008059
- Molecular Weight:183.25 g/mol
- Boiling Pt:294…296 °C
- Melting Pt:41579 °C
- Density:1.063 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:202-032-2
- UN:2735
- ADR:8,II
3 Options Available Below
Product Details & Documents
Synonyms: α-Phenylbenzylamine, α-Aminodiphenylmethane, Diphenylmethanamine, Aminodiphenylmethane, 1,1-Diphenylmethanamine, Benzhydrylamin
- CAS Number:91-00-9
- Formula:C₁₃H₁₃N
- MDL Number:MFCD00008059
- Molecular Weight:183.25 g/mol
- Boiling Pt:294…296 °C
- Melting Pt:41579 °C
- Density:1.063 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:202-032-2
- UN:2735
- ADR:8,II



