
1,3-Dimethoxy-4-iodobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:20469-63-0
:
Formula:C₈H₉IO₂
MDL Number:MFCD00059268
:264.06 g/mol
Boiling Pt:135 °C/14mmHg
Melting Pt:37…41 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient