Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amino-1,3,5-trimethyl-1H-pyrazole
4-Amino-1,3,5-trimethyl-1H-pyrazole
4-Amino-1,3,5-trimethyl-1H-pyrazole
Irritant
4-Amino-1,3,5-trimethyl-1H-pyrazole
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Amino-1,3,5-trimethyl-1H-pyrazole
Supplier:
Apollo Scientific
About this item
CAS Number:
28466-21-9
:
Formula:
C₆H₁₁N₃
MDL Number:
MFCD00052883
:
125.17 g/mol
Melting Pt:
99…105 °C
:
GHS07
Storage Temperature:
Ambient