Order Entry
Export
ContactUsLinkComponent
 
5-Chloro-2-methoxybenzoic acid 97%
Catalog # APOSOR8055-500G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-2-methoxybenzoic acid 97%
Catalog # APOSOR8055-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :186.59 g/mol
Melting Pt:98…100 °C
EINECS:222-343-7
 :GHS07
Storage Temperature:Ambient
CAS Number:3438-16-2
Formula:C₈H₇ClO₃
MDL Number:MFCD00017546