Order Entry
Export
ContactUsLinkComponent
5-(2-Bromoethoxy)-2,3-dihydro-1,4-benzodioxine 97%
Irritant
5-(2-Bromoethoxy)-2,3-dihydro-1,4-benzodioxine 97%
Catalog # APOSOR8037-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-(2-Bromoethoxy)-2,3-dihydro-1,4-benzodioxine 97%
Catalog # APOSOR8037-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.11 g/mol
Melting Pt:85…86 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1710-62-9
Formula:C₁₀H₁₁BrO₃
MDL Number:MFCD00099411