
1,2-Dichloro-4-methyl-5-(methylsulphonyl)-3-nitrobenzene
Catalog # APOSOR7981-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:284.12 g/mol
Melting Pt:163…165 °C
:GHS07
Storage Temperature:Ambient
CAS Number:849035-79-6
Formula:C₈H₇Cl₂NO₄S
MDL Number:MFCD04037949