Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dichlorobenzaldehyde

2,6-Dichlorobenzaldehyde

Bottle, 100 g
  APOSOR7973-100G
Corrosive
2,6-Dichlorobenzaldehyde
  APOSOR7973-100G
 :  83-38-5
2,6-Dichlorobenzaldehyde

Bottle, 100 g

 :  83-38-5
 
21,80 €
Log in for availability
1 * 100 g
 
  • CAS Number:83-38-5
  • Formula:C₇H₄Cl₂O
  • MDL Number:MFCD00003307
  • Molecular Weight:175.01 g/mol
  • Boiling Pt:231 °C/8mmHg
  • Melting Pt:68…73 °C
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:201-472-2
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications