Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Xylose

D-(+)-Xylose

Irritant
Supplier:   Apollo Scientific
D-(+)-Xylose
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:58-86-6
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00151475
  • Molecular Weight:150.13 g/mol
  • Melting Pt:147…158 °C
  • Density:1.525 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:200-400-7

Documents

Product Family Options

Product Information