

2,4-Dimethylthiophenol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13616-82-5
:
Formula:C₈H₁₀S
MDL Number:MFCD00010019
:138.23 g/mol
Boiling Pt:207…208 °C
Density:1.022 g/cm³ (25 °C)
Flash Pt:58 °C
Storage Temperature:Refrigerator
EINECS:237-100-0
UN:1993
ADR:3,III
:GHS02
:GHS07