Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dichloroimidazole
4,5-Dichloroimidazole
4,5-Dichloroimidazole
Irritant
4,5-Dichloroimidazole
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,5-Dichloroimidazole
Supplier:
Apollo Scientific
About this item
CAS Number:
15965-30-7
:
Formula:
C₃H₂Cl₂N₂
MDL Number:
MFCD00005195
:
136.97 g/mol
Melting Pt:
181…185 °C
:
GHS07
Storage Temperature:
Ambient