
1-(4,6-Dimethylpyrimidin-2-yl)piperidine-4-carboxylic acid
Catalog # APOSOR7849-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.29 g/mol
Melting Pt:158…160 °C
:GHS07
Storage Temperature:Ambient
CAS Number:797028-97-8
Formula:C₁₂H₁₇N₃O₂
MDL Number:MFCD03426079