Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(5-Phenyl-1,3,4-oxadiazol-2-yl)piperidine
4-(5-Phenyl-1,3,4-oxadiazol-2-yl)piperidine 90+%
4-(5-Phenyl-1,3,4-oxadiazol-2-yl)piperidine 90+%
APOSOR7806-250MG
:
Apollo Scientific
:
4-(5-Phenyl-1,3,4-oxadiazol-2-yl)piperidine 90+%
APOSOR7806-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
229.28 g/mol
:
89…91 °C
:
GHS07
:
Ambient
:
280110-78-3
:
C₁₃H₁₅N₃O
:
MFCD03426083