Methyl-3-[(4,6-dimethylpyrimidin-2-yl)amino]thiophene-2-carboxylate 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:388565-75-1
:
Formula:C₁₂H₁₃N₃O₂S
MDL Number:MFCD00202934
:263.32 g/mol
Melting Pt:119…121 °C
Storage Temperature:Ambient