
1-(4-Chlorophenyl)-1,2-propandione 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10557-21-8
:
Formula:C₉H₇ClO₂
MDL Number:MFCD01941300
:182.61 g/mol
Boiling Pt:60…65 °C/0.02mmHg
Melting Pt:30…32 °C
:GHS07
Storage Temperature:Ambient