Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-1,8-naphthalic anhydride
4-Bromo-1,8-naphthalic anhydride
4-Bromo-1,8-naphthalic anhydride
Irritant
4-Bromo-1,8-naphthalic anhydride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Bromo-1,8-naphthalic anhydride
Supplier:
Apollo Scientific
About this item
CAS Number:
81-86-7
:
Formula:
C₁₂H₅BrO₃
MDL Number:
MFCD00006927
:
277.0703 g/mol
Melting Pt:
217…219 °C
EINECS:
201-382-3
:
GHS07
Storage Temperature:
Ambient