2,4-Dinitrobenzaldehyde
Bottle, 5 g
Catalog # APOSOR7733-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:528-75-6
- Formula:C₇H₄N₂O₅
- MDL Number:MFCD00013376
- Molecular Weight:196.12 g/mol
- Boiling Pt:190 °C
- Melting Pt:66…70 °C
- Storage Temperature:Ambient
- EINECS:208-440-7
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g



