Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dimethoxy-2-(piperidin-4-yl)pyrimidine
4,6-Dimethoxy-2-(piperidin-4-yl)pyrimidine 95+%
4,6-Dimethoxy-2-(piperidin-4-yl)pyrimidine 95+%
:
Apollo Scientific
4,6-Dimethoxy-2-(piperidin-4-yl)pyrimidine 95+%
:
Apollo Scientific
:
849924-99-8
:
:
C₁₁H₁₇N₃O₂
:
MFCD03406618
:
223.28 g/mol
:
48…51 °C
:
GHS07
:
Ambient