

1-(5-Bromopyrimidin-2-yl)-[1,4]-diazepane 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:849021-44-9
:
Formula:C₉H₁₃BrN₄
MDL Number:MFCD03426069
:257.14 g/mol
Melting Pt:203…207 °C
:GHS05
:GHS07
Storage Temperature:Ambient