Order Entry
Export
ContactUsLinkComponent
 
Irritant
2-(1,8-Naphthyridin-2-yl)phenol 95+%
Catalog # APOSOR7617-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(1,8-Naphthyridin-2-yl)phenol 95+%
Catalog # APOSOR7617-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:193…196 °C
 :222.25 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:65182-56-1
Formula:C₁₄H₁₀N₂O
MDL Number:MFCD00203050