Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Chloro[1,1'-biphenyl]-2-carbaldehyde
2'-Chloro[1,1'-biphenyl]-2-carbaldehyde
2'-Chloro[1,1'-biphenyl]-2-carbaldehyde
Irritant
2'-Chloro[1,1'-biphenyl]-2-carbaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2'-Chloro[1,1'-biphenyl]-2-carbaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
223575-76-6
:
Formula:
C₁₃H₉ClO
MDL Number:
MFCD04039056
:
216.67 g/mol
:
GHS07
Storage Temperature:
Ambient