Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro-4-methoxyphenylboronic acid
3,5-Dichloro-4-methoxyphenylboronic acid
3,5-Dichloro-4-methoxyphenylboronic acid
Irritant
3,5-Dichloro-4-methoxyphenylboronic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Dichloro-4-methoxyphenylboronic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
175883-61-1
:
Formula:
C₇H₇BCl₂O₃
MDL Number:
MFCD04039018
:
220.8500 g/mol
:
GHS07
Storage Temperature:
Refrigerator