Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-N-Boc-1,4-cyclohexanediamine
trans-N-Boc-1,4-cyclohexanediamine
trans-N-Boc-1,4-cyclohexanediamine
Irritant
trans-N-Boc-1,4-cyclohexanediamine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
trans-N-Boc-1,4-cyclohexanediamine
Supplier:
Apollo Scientific
About this item
CAS Number:
177906-48-8
:
Formula:
C₁₁H₂₂N₂O₂
MDL Number:
MFCD03001719
:
214.30 g/mol
Melting Pt:
74.5…75.5 °C
:
GHS07
Storage Temperature:
Ambient