Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromotetrazolo[1,5-a]pyridine
6-Bromotetrazolo[1,5-a]pyridine 98%
6-Bromotetrazolo[1,5-a]pyridine 98%
:
Apollo Scientific
6-Bromotetrazolo[1,5-a]pyridine 98%
:
Apollo Scientific
:
35235-74-6
:
:
C₅H₃BrN₄
:
MFCD09258642
:
148…153 °C
:
GHS07
:
Refrigerator
Environmentally Preferable: