Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Methyl-1,3-cyclohexanedione
5-Methyl-1,3-cyclohexanedione 98%
5-Methyl-1,3-cyclohexanedione 98%
Irritant
5-Methyl-1,3-cyclohexanedione 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Methyl-1,3-cyclohexanedione 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
4341-24-6
:
Formula:
C₇H₁₀O₂
MDL Number:
MFCD00010379
:
126.16 g/mol
Melting Pt:
125…132 °C
:
GHS07
Storage Temperature:
Ambient