Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxy-2-methylpropiophenone
2-Hydroxy-2-methylpropiophenone 98%
2-Hydroxy-2-methylpropiophenone 98%
:
Apollo Scientific
2-Hydroxy-2-methylpropiophenone 98%
:
Apollo Scientific
:
7473-98-5
:
:
C₁₀H₁₂O₂
:
MFCD00040710
:
164.2000 g/mol
:
102…103 °C/3.75mmHg
:
1.077 g/cm³ (25 °C)
:
122 °C
:
GHS07
:
Ambient