Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dibromobenzaldehyde
2,5-Dibromobenzaldehyde
2,5-Dibromobenzaldehyde
:
Apollo Scientific
2,5-Dibromobenzaldehyde
:
Apollo Scientific
:
74553-29-0
:
:
C₇H₄Br₂O
:
MFCD00462864
:
263.92 g/mol
:
88…92 °C
:
2923
:
8+6.1,II
:
GHS05
:
GHS06
:
GHS09
:
Ambient