Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Phenylenediacryloyl chloride
1,4-Phenylenediacryloyl chloride
1,4-Phenylenediacryloyl chloride
:
Apollo Scientific
1,4-Phenylenediacryloyl chloride
:
Apollo Scientific
:
35288-49-4
:
:
C₁₂H₈Cl₂O₂
:
MFCD01109676
:
255.10 g/mol
:
171…172 °C
:
Refrigerator
:
1759
:
8,III
:
GHS05