1,3-Propanedithiol 97%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1,3-Dimercaptopropane
- CAS Number:109-80-8
- Formula:C₃H₈S₂
- MDL Number:MFCD00004904
- Molecular Weight:108.23 g/mol
- Boiling Pt:168…170 °C
- Melting Pt:–79 °C
- Density:1.077 g/cm³ (25 °C)
- Flash Pt:63 °C
- Storage Temperature:Refrigerator
- UN:3334
- ADR:9,III
5 Options Available Below
Product Details & Documents
Synonyms: 1,3-Dimercaptopropane
- CAS Number:109-80-8
- Formula:C₃H₈S₂
- MDL Number:MFCD00004904
- Molecular Weight:108.23 g/mol
- Boiling Pt:168…170 °C
- Melting Pt:–79 °C
- Density:1.077 g/cm³ (25 °C)
- Flash Pt:63 °C
- Storage Temperature:Refrigerator
- UN:3334
- ADR:9,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



