

2,5-Dibromo-p-xylene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1074-24-4
:
Formula:C₈H₈Br₂
MDL Number:MFCD00000091
:263.96 g/mol
Boiling Pt:261 °C
Melting Pt:72…77 °C
Storage Temperature:Ambient
EINECS:214-038-2
UN:3077
ADR:9+EHS,III
:GHS07
:GHS09