
1,4-Dioxa-8-azaspiro[4.5]decane 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:177-11-7
:
Formula:C₇H₁₃NO₂
MDL Number:MFCD00005976
:143.186 g/mol
Boiling Pt:99…100 °C/14mmHg
Density:1.17 g/cm³ (25 °C)
Flash Pt:81 °C
EINECS:205-868-6
:GHS07
Storage Temperature:Ambient