
1-Cyclopentylpiperazine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21043-40-3
:
Formula:C₉H₁₈N₂
MDL Number:MFCD00168029
:154.26 g/mol
Boiling Pt:103…105 °C/11mmHg
Melting Pt:25 °C
Density:0.97 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient