Order Entry
Export
ContactUsLinkComponent
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid 95+%
Irritant
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid 95+%
Catalog # APOSOR6889-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-(4,6-Dimethylpyrimidin-2-yl)-N-methyl-4-aminobenzoic acid 95+%
Catalog # APOSOR6889-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:172…174 °C
 :257.29 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:387350-50-7
Formula:C₁₄H₁₅N₃O₂
MDL Number:MFCD00203268