Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3,5-dichlorobenzoate
Methyl-3,5-dichlorobenzoate 98%
Methyl-3,5-dichlorobenzoate 98%
Methyl-3,5-dichlorobenzoate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-3,5-dichlorobenzoate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
2905-67-1
:
Formula:
C₈H₆Cl₂O₂
MDL Number:
MFCD00032710
:
205.04 g/mol
Storage Temperature:
Ambient