0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1-Methyl-2-formylbenzimidazole
1-Methyl-2-formylbenzimidazole
1-Methyl-2-formylbenzimidazole
APOSOR6737-1G
:
Apollo Scientific
:
1-Methyl-2-formylbenzimidazole
APOSOR6737-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
160.176 g/mol
:
118…121 °C
:
GHS07
:
Ambient
:
3012-80-4
:
C₉H₈N₂O
:
MFCD00142655