Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-benzodioxole-5-sulphonyl chloride
1,3-benzodioxole-5-sulphonyl chloride 95+%
1,3-benzodioxole-5-sulphonyl chloride 95+%
:
Apollo Scientific
1,3-benzodioxole-5-sulphonyl chloride 95+%
:
Apollo Scientific
:
115010-10-1
:
:
C₇H₅ClO₄S
:
MFCD04974524
:
220.623 g/mol
:
48…51 °C
:
3261
:
8,III
:
GHS05
:
GHS07
:
Ambient