Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dimethylbenzimidazole
5,6-Dimethylbenzimidazole
5,6-Dimethylbenzimidazole
Irritant
5,6-Dimethylbenzimidazole
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5,6-Dimethylbenzimidazole
Supplier:
Apollo Scientific
About this item
CAS Number:
582-60-5
:
Formula:
C₉H₁₀N₂
MDL Number:
MFCD00005603
:
146.19 g/mol
Melting Pt:
202…206 °C
EINECS:
209-488-1
:
GHS07
Storage Temperature:
Ambient