Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2,4-dimethoxypyrimidine
5-Bromo-2,4-dimethoxypyrimidine
5-Bromo-2,4-dimethoxypyrimidine
Irritant
5-Bromo-2,4-dimethoxypyrimidine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Bromo-2,4-dimethoxypyrimidine
Supplier:
Apollo Scientific
About this item
CAS Number:
56686-16-9
:
Formula:
C₆H₇BrN₂O₂
MDL Number:
MFCD00038016
:
219.0400 g/mol
Melting Pt:
62…65 °C
:
GHS07
Storage Temperature:
Ambient