Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-2,4-dinitrotoluene
5-Chloro-2,4-dinitrotoluene
5-Chloro-2,4-dinitrotoluene
Irritant
5-Chloro-2,4-dinitrotoluene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Chloro-2,4-dinitrotoluene
Supplier:
Apollo Scientific
About this item
CAS Number:
51676-74-5
:
Formula:
C₇H₅ClN₂O₄
MDL Number:
MFCD00041235
:
216.58 g/mol
Melting Pt:
87…91 °C
:
GHS07
Storage Temperature:
Ambient