Order Entry
Export
Orders LinkContactUsLinkComponent
9-Hydroxyfluorene 97%

9-Hydroxyfluorene 97%

Bottle, 25 g
Catalog # APOSOR6505-25G
Irritant
Supplier:   Apollo Scientific
9-Hydroxyfluorene 97%
Catalog # APOSOR6505-25G
Supplier:  Apollo Scientific
CAS Number:  1689-64-1
9-Hydroxyfluorene 97%

Bottle, 25 g

CAS Number:  1689-64-1
Price
102,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  9-Fluorenol, Fluoren-9-ol
  • CAS Number:1689-64-1
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00001135
  • Molecular Weight:182.22 g/mol
  • Melting Pt:155…157 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications