Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Dicyanomethylene)indan-1,3-dione
2-(Dicyanomethylene)indan-1,3-dione 98%
2-(Dicyanomethylene)indan-1,3-dione 98%
:
Apollo Scientific
2-(Dicyanomethylene)indan-1,3-dione 98%
:
Apollo Scientific
:
16954-74-8
:
C₁₂H₄N₂O₂
:
MFCD00051930
:
208.18 g/mol
:
270 °C (dec.)
:
Ambient
:
3439
:
6.1,III
:
GHS07