Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dibromothiazole
2,4-Dibromothiazole
2,4-Dibromothiazole
Corrosive
Irritant
2,4-Dibromothiazole
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
2,4-Dibromothiazole
Supplier:
Apollo Scientific
About this item
CAS Number:
4175-77-3
:
Formula:
C₃HBr₂NS
MDL Number:
MFCD01318994
:
242.92 g/mol
Melting Pt:
80…84 °C
:
GHS05
:
GHS07
Storage Temperature:
Ambient