2,5-Diaminopyridine dihydrochloride 96%
Catalog # APOSOR6458-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:182.05 g/mol
Melting Pt:264 °C
Storage Temperature:Ambient
CAS Number:26878-35-3
Formula:C₅H₉Cl₂N₃
MDL Number:MFCD00040818