2,6-Pyridinedimethanol
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,6-Bis(hydroxymethyl)pyridine
- CAS Number:1195-59-1
- Formula:C₇H₉NO₂
- MDL Number:MFCD00006351
- Molecular Weight:139.15 g/mol
- Boiling Pt:185 °C/15mmHg
- Melting Pt:110…115 °C
- Storage Temperature:Ambient
- EINECS:214-803-0
3 Options Available Below
Product Details & Documents
Synonyms: 2,6-Bis(hydroxymethyl)pyridine
- CAS Number:1195-59-1
- Formula:C₇H₉NO₂
- MDL Number:MFCD00006351
- Molecular Weight:139.15 g/mol
- Boiling Pt:185 °C/15mmHg
- Melting Pt:110…115 °C
- Storage Temperature:Ambient
- EINECS:214-803-0



