

3-Bromo-2,6-dimethylpyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3430-31-7
:
Formula:C₇H₈BrN
MDL Number:MFCD00235164
:186.0491 g/mol
Boiling Pt:56…58 °C
Density:1.431 g/cm³ (25 °C)
Flash Pt:86.7 °C
:GHS05
:GHS07
Storage Temperature:Ambient