
1-(Bromomethyl)-2,3-dimethylbenzene 95%
Catalog # APOSOR6439-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.0876 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:81093-21-2
Formula:C₉H₁₁Br
MDL Number:MFCD03844741