0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
7-Bromopyrido[3,2-b]pyrazine
7-Bromopyrido[3,2-b]pyrazine 97%
7-Bromopyrido[3,2-b]pyrazine 97%
Irritant
7-Bromopyrido[3,2-b]pyrazine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
7-Bromopyrido[3,2-b]pyrazine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
52333-42-3
:
Formula:
C₇H₄BRN₃
MDL Number:
MFCD01105748
:
210.03 g/mol
:
GHS07
Storage Temperature:
Ambient