2,3-Dimethoxypyridine 96%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:52605-97-7
- Formula:C₇H₉NO₂
- MDL Number:MFCD03095043
- Molecular Weight:139.15 g/mol
- Storage Temperature:Ambient
5 Options Available Below
Product Details & Documents
- CAS Number:52605-97-7
- Formula:C₇H₉NO₂
- MDL Number:MFCD03095043
- Molecular Weight:139.15 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



