3-Nitrophthalic anhydride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:641-70-3
:
Formula:C₈H₃NO₅
MDL Number:MFCD00005921
:193.12 g/mol
Melting Pt:163…165 °C
EINECS:211-373-6
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: