
4,6-Dichloro-o-toluidine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:30273-00-8
:
Formula:C₇H₇Cl₂N
MDL Number:MFCD00044074
:176.05 g/mol
Boiling Pt:82…84 °C/0.1mmHg
Melting Pt:42…46 °C
:GHS07
Storage Temperature:Ambient