
5-Bromo-6-oxo-1,6-dihydro-3-pyridine carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:41668-13-7
:
Formula:C₆H₄BrNO₃
MDL Number:MFCD03791237
:218.01 g/mol
Melting Pt:334…336 °C
:GHS07
Storage Temperature:Ambient